Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JPX SIRNA Assays ›
          Gene Symbol:
          JPX
          Gene Name:
          JPX transcript, XIST activator (non-protein coding)
          Gene Aliases:
          DCBALD06, ENOX, LINC00183, NCRNA00183
          Chromosome Location:
          Chr.X: 73944324 - 74070384 on Build GRCh38
          UniGene ID:
          Hs.648316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n504762

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JPX
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024582.1
          2 185
          GenBank mRNA AK056172.1 -
          2 167
          AV714079.1 - LIT1011
          Jpx
          170
          BC071776.1 -
          2 181
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA979750.1 - 2 158 Chr. X - 73944351 - 73947352 [+] on Build GRCh38
          GenBankmRNA BG504771.1 - 2 164 Chr. X - 73944346 - 73947528 [+] on Build GRCh38
          GenBankmRNA BP872745.1 - 2 164 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BM548256.1 - 217 Chr. - - [-] on Build
          GenBankmRNA BP871836.1 - 2 163 Chr. X - 73944346 - 73947297 [+] on Build GRCh38
          GenBankmRNA CN270680.1 - 2 150 Chr. X - 73944359 - 73999372 [+] on Build GRCh38
          GenBankmRNA AW963219.1 - 157 Chr. - - [-] on Build
          GenBankmRNA AV705096.1 - 2 120 Chr. X - 73944389 - 74004413 [+] on Build GRCh38
          GenBankmRNA BU934969.1 - 2 165 Chr. X - 73944346 - 73999254 [+] on Build GRCh38
          GenBankmRNA Z44786.1 - 152 Chr. X - 123628779 - 123628794 [-] on Build GRCh38
          GenBankmRNA AL711317.1 - 2 184 Chr. X - 73944324 - 73999368 [+] on Build GRCh38
          GenBankmRNA BG612854.1 - 2 166 Chr. X - 73944343 - 73999478 [+] on Build GRCh38
          GenBankmRNA CB993733.1 - 194 Chr. - - [-] on Build
          GenBankmRNA HY065667.1 - 2 171 Chr. X - 73944340 - 74010499 [+] on Build GRCh38
          GenBankmRNA BP872499.1 - 2 185 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP873595.1 - 2 167 Chr. X - 73944342 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP872256.1 - 2 164 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DB255033.1 - 2 172 Chr. X - 73944339 - 73947366 [+] on Build GRCh38
          GenBankmRNA CF129475.1 - 2 150 Chr. X - 73944359 - 74004553 [+] on Build GRCh38
          GenBankmRNA F12346.1 - 126 Chr. - - [-] on Build
          GenBankmRNA DA106100.1 - 2 167 Chr. X - 73944342 - 74004417 [+] on Build GRCh38
          GenBankmRNA DA363254.1 - 2 184 Chr. X - 73944326 - 73997077 [+] on Build GRCh38
          GenBankmRNA CN263834.1 - 2 153 Chr. X - 73944356 - 73999314 [+] on Build GRCh38
          GENCODE ENST00000414209.5 - 2 167 Chr. X - 73944342 - 74000491 [+] on Build GRCh38
          GenBankmRNA HY103315.1 - 2 167 Chr. X - 73944342 - 73947268 [+] on Build GRCh38
          GENCODE ENST00000453317.1 - 2 163 Chr. X - 73944346 - 73944946 [+] on Build GRCh38
          GenBankmRNA BM810643.1 - 140 Chr. - - [-] on Build
          GenBankmRNA BU164379.1 - 2 160 Chr. X - 73944349 - 73998832 [+] on Build GRCh38
          GenBankmRNA BP871739.1 - 2 164 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP872588.1 - 2 164 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP871919.1 - 2 165 Chr. X - 73944344 - 73999613 [+] on Build GRCh38
          GenBankmRNA DA841100.1 - 2 164 Chr. X - 73944345 - 73947361 [+] on Build GRCh38
          GenBankmRNA BG613915.1 - 2 168 Chr. X - 73944342 - 73999860 [+] on Build GRCh38
          GenBankmRNA AL708895.1 - 2 172 Chr. X - 73944338 - 73947333 [+] on Build GRCh38
          GenBankmRNA BU537716.1 - 2 179 Chr. X - 73944333 - 73947334 [+] on Build GRCh38
          GenBankmRNA BP871940.1 - 2 164 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DA331725.1 - 2 167 Chr. X - 73944342 - 74004540 [+] on Build GRCh38
          GenBankmRNA BP873466.1 - 2 185 Chr. X - 73944324 - 73999111 [+] on Build GRCh38
          GENCODE ENST00000602294.5 - 2 185 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP871783.1 - 2 164 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA R19849.1 - 2 148 Chr. X - 73944385 - 74047792 [+] on Build GRCh38
          GenBankmRNA HY088843.1 - 2 171 Chr. X - 73944340 - 74010567 [+] on Build GRCh38
          GenBankmRNA HY172225.1 - 2 168 Chr. X - 73944342 - 73999019 [+] on Build GRCh38
          GenBankmRNA BP872277.1 - 2 185 Chr. X - 73944324 - 73999592 [+] on Build GRCh38
          GenBankmRNA DA103086.1 - 2 166 Chr. X - 73944343 - 74004499 [+] on Build GRCh38
          GenBankmRNA BG435566.1 - 2 182 Chr. X - 73944328 - 74069325 [+] on Build GRCh38
          GenBankmRNA BU589636.1 - 2 166 Chr. X - 73944346 - 73944948 [+] on Build GRCh38
          GenBankmRNA DA427047.1 - 2 183 Chr. X - 73944326 - 73947342 [+] on Build GRCh38
          GenBankmRNA CB991754.1 - 194 Chr. - - [-] on Build
          GenBankmRNA DB240432.1 - 2 163 Chr. X - 73944346 - 73947385 [+] on Build GRCh38
          GenBankmRNA DA881370.1 - 2 162 Chr. X - 73944347 - 73999014 [+] on Build GRCh38
          GenBankmRNA DA679611.1 - 2 167 Chr. X - 73944342 - 73999105 [+] on Build GRCh38
          GenBankmRNA C16093.1 - 1 103 Chr. 3 - 71682388 - 71682656 [-] on Build GRCh38
          GenBankmRNA DA731241.1 - 2 183 Chr. X - 73944326 - 73998845 [+] on Build GRCh38
          GenBankmRNA BP873513.1 - 2 164 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA T32551.1 - 156 Chr. 15 - 42410703 - 42410722 [-] on Build GRCh38
          GenBankmRNA BP872419.1 - 2 185 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BF686802.1 - 148 Chr. - - [-] on Build
          GenBankmRNA DA700446.1 - 2 167 Chr. X - 73944342 - 73947318 [+] on Build GRCh38
          GenBankmRNA DB250056.1 - 2 163 Chr. X - 73944346 - 73999096 [+] on Build GRCh38
          GenBankmRNA BG427198.1 - 2 168 Chr. X - 73944342 - 73995436 [+] on Build GRCh38
          GenBankmRNA DW450219.1 - 2 180 Chr. X - 73944342 - 73944645 [+] on Build GRCh38
          GenBankmRNA DW466961.1 - 2 182 Chr. X - 73944340 - 73994872 [+] on Build GRCh38
          GenBankmRNA BP871862.1 - 2 169 Chr. X - 73944340 - 73947374 [+] on Build GRCh38
          GenBankmRNA BP873504.1 - 2 161 Chr. X - 73944356 - 74004351 [+] on Build GRCh38
          GENCODE ENST00000602772.5 - 2 181 Chr. X - 73944328 - 74070408 [+] on Build GRCh38
          GenBankmRNA BP872587.1 - 2 165 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA HY081032.1 - 2 170 Chr. X - 73944339 - 73947244 [+] on Build GRCh38
          GENCODE ENST00000602938.5 - 2 185 Chr. X - 73944324 - 73999114 [+] on Build GRCh38
          GenBankmRNA BP873185.1 - 2 165 Chr. X - 73944344 - 74004355 [+] on Build GRCh38
          GenBankmRNA H84526.1 - 145 Chr. - - [-] on Build
          GenBankmRNA BU599790.1 - 2 170 Chr. X - 73944340 - 74004542 [+] on Build GRCh38
          GenBankmRNA BP872964.1 - 2 164 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BF797301.1 - 2 152 Chr. X - 73944357 - 73999368 [+] on Build GRCh38
          GenBankmRNA R27273.1 - 149 Chr. - - [-] on Build
          GenBankmRNA BP872836.1 - 2 165 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA DB210310.1 - 2 178 Chr. X - 73944332 - 73999117 [+] on Build GRCh38
          GenBankmRNA DA711535.1 - 2 168 Chr. X - 73944342 - 73947373 [+] on Build GRCh38
          GenBankmRNA CB998281.1 - 194 Chr. - - [-] on Build
          GENCODE ENST00000602920.5 - 2 185 Chr. X - 73944324 - 74004551 [+] on Build GRCh38
          GenBankmRNA BP872490.1 - 2 165 Chr. X - 73944344 - 74004345 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0