Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: LINC00472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026807.2 | 4 | 2170 | |||
NR_121612.1 | 4 | 2221 | ||||
NR_121613.1 | 3 | 2155 | ||||
NR_121614.1 | 3 | 2104 | ||||
GenBank mRNA | AK023251.1 | - | 4 | 2170 | ||
BM720568.1 | - | 1 | 570 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000426635.6 | - | 4 | 2194 | Chr. 6 - 71414445 - 71420769 [-] on Build GRCh38 |