Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CASC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103848.1 | 3 | 505 | |||
NR_103849.2 | 2 | 453 | ||||
NR_103850.2 | 2 | 401 | ||||
GenBank mRNA | BE866214.1 | - | 2 | 381 | ||
CN273839.1 | - | 2 | 424 | |||
CN418510.1 | - | 2 | 374 | |||
CX870402.1 | - | 2 | 453 |