Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLEU1 SIRNA Assays ›
          Gene Symbol:
          DLEU1
          Gene Name:
          deleted in lymphocytic leukemia 1 (non-protein coding)
          Gene Aliases:
          BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6
          Chromosome Location:
          Chr.13: 50082169 - 50528643 on Build GRCh38
          UniGene ID:
          Hs.591229
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n541183

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLEU1
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_109974.1
          4 552
          GenBank mRNA AJ412026.1 - LIT1926
          BCMS
          3 516
          AJ412027.1 - LIT1925
          BCMS
          3 223
          AJ412029.1 - LIT1923
          BCMS
          3 223
          AJ412031.1 - LIT1917
          BCMS
          8 1027
          AJ412033.1 - LIT1912
          BCMS
          2 129
          AJ412034.1 - LIT1911
          BCMS
          2 129
          AJ412036.1 - LIT1909
          BCMS
          2 129
          AJ412037.1 - LIT1908
          BCMS
          2 129
          AJ412042.1 - LIT1903
          BCMS
          3 301
          AJ412043.1 - LIT1902
          BCMS
          4 414
          AJ412054.1 - LIT1886
          BCMS
          5 684
          AJ412055.1 - LIT1885
          BCMS
          4 550
          AJ412056.1 - LIT1884
          BCMS
          4 615
          AJ412057.1 - LIT1876
          BCMS
          5 663
          AJ412058.1 - LIT1875
          BCMS
          4 569
          AJ412059.1 - LIT1874
          BCMS
          5 663
          AJ412060.1 - LIT1873
          BCMS
          3 1649
          AJ412061.1 - LIT1872
          BCMS
          5 663
          AJ412062.1 - LIT1871
          BCMS
          5 663
          AJ412063.1 - LIT1870
          BCMS
          2 110
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GENCODE ENST00000472136.5 - 2 130 Chr. 13 - 50415089 - 50495469 [+] on Build GRCh38
          GENCODE ENST00000475913.5 - 3 301 Chr. 13 - 50185880 - 50492761 [+] on Build GRCh38
          GENCODE ENST00000461527.6 - 5 684 Chr. 13 - 50082171 - 50492761 [+] on Build GRCh38
          GENCODE ENST00000498557.5 - 2 130 Chr. 13 - 50415089 - 50475566 [+] on Build GRCh38
          GENCODE ENST00000468168.5 - 4 550 Chr. 13 - 50082171 - 50528064 [+] on Build GRCh38
          GENCODE ENST00000491482.5 - 2 110 Chr. 13 - 50390198 - 50528064 [+] on Build GRCh38
          GENCODE ENST00000463474.6 - 4 615 Chr. 13 - 50082171 - 50528064 [+] on Build GRCh38
          GenBankmRNA AJ412003.3 - 1 16 Chr. 13 - 50433413 - 50433550 [+] on Build GRCh38
          GENCODE ENST00000462427.1 - 2 130 Chr. 13 - 50415089 - 50533861 [+] on Build GRCh38
          GENCODE ENST00000460525.5 - 2 130 Chr. 13 - 50415089 - 50533861 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0