Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120656.1 | 4 | 468 | |||
NR_120657.1 | 3 | 362 | ||||
NR_120658.1 | 3 | 358 | ||||
NR_120659.1 | 4 | 479 | ||||
GenBank mRNA | AK123067.1 | - | 4 | 468 | ||
DB446959.1 | - | 3 | 330 | |||
DB454367.1 | - | 4 | 425 | |||
HY051619.1 | - | 3 | 308 |