Search
Search

ITGB2-AS1| Gene Symbol: ITGB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038311.1 | 5 | 1139 | |||
| NR_038312.1 | 4 | 1096 | ||||
| NR_038313.1 | 5 | 1115 | ||||
| NR_038314.1 | 4 | 1044 | ||||
| NR_038315.1 | 4 | 1040 | ||||
| NR_038316.1 | 3 | 1001 | ||||
| GenBank mRNA | BC040064.1 | - | 4 | 1028 | ||