Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC101927179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125880.1 | 1 | 95 | |||
NR_125881.1 | 1 | 95 | ||||
GenBank mRNA | AL599247.1 | - | 1 | 96 | ||
BX280044.1 | - | 1 | 57 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | W58114.1 | - | 58 | Chr. - - [-] on Build |