Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFAS1 SIRNA Assays ›
          Gene Symbol:
          ZFAS1
          Gene Name:
          ZNFX1 antisense RNA 1
          Gene Aliases:
          C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1
          Chromosome Location:
          Chr.20: 49278178 - 49289260 on Build GRCh38
          UniGene ID:
          Hs.356766
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n548942

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFAS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003604.2 5 944
          NR_003605.1 5 625
          NR_003606.2 3 796
          NR_036658.1 4 882
          NR_036659.1 4 440
          GenBank mRNA AK124175.1 - 1 2590
          AY339614.1 - 5 446
          AY513722.1 - 5 944
          AY513723.1 - 5 625
          BC032480.1 - 3 315
          BC042807.1 - 5 484
          BC060315.1 - 4 405
          BC062222.1 - 4 444
          BC062447.1 - 3 272
          BC105282.1 - 5 484
          CB137861.1 - 3 332
          CR749730.1 - 1 2097
          CR936805.1 - 4 8564
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD385315.1 - 434 Chr. - - [-] on Build
          GenBankmRNA BG260310.1 - 306 Chr. - - [-] on Build
          GenBankmRNA CB155482.1 - 6 548 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA CF123544.1 - 3 372 Chr. 20 - 49278639 - 49289257 [+] on Build GRCh38
          GenBankmRNA DA313134.1 - 5 481 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA BU954336.1 - 5 483 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW954155.1 - 448 Chr. - - [-] on Build
          GenBankmRNA HY104293.1 - 4 417 Chr. 20 - 49278643 - 49289213 [+] on Build GRCh38
          GenBankmRNA CB151494.1 - 4 419 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU940503.1 - 5 485 Chr. 20 - 49278642 - 49289253 [+] on Build GRCh38
          GenBankmRNA BU849377.1 - 4 421 Chr. 20 - 49278642 - 49289232 [+] on Build GRCh38
          GenBankmRNA BI545004.1 - 5 489 Chr. 20 - 49278638 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295715.1 - 5 471 Chr. 20 - 49278651 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG109475.1 - 5 451 Chr. 20 - 49278669 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM782711.1 - 4 418 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA AW953042.1 - 408 Chr. - - [-] on Build
          GenBankmRNA BE897330.1 - 4 398 Chr. 20 - 49278662 - 49289257 [+] on Build GRCh38
          GenBankmRNA CD674657.1 - 5 463 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY128835.1 - 5 479 Chr. 20 - 49278643 - 49289222 [+] on Build GRCh38
          GenBankmRNA BU599962.1 - 484 Chr. - - [-] on Build
          GenBankmRNA BM701753.1 - 5 440 Chr. 20 - 49278682 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU927839.1 - 421 Chr. - - [-] on Build
          GenBankmRNA CD248194.1 - 5 498 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BJ995977.1 - 4 418 Chr. 20 - 49278642 - 49289233 [+] on Build GRCh38
          GenBankmRNA CN353461.1 - 4 397 Chr. 20 - 49278663 - 49289231 [+] on Build GRCh38
          GenBankmRNA AA484881.1 - 4 414 Chr. 20 - 49278653 - 49289218 [+] on Build GRCh38
          GenBankmRNA BG110424.1 - 4 450 Chr. 20 - 49278645 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG112759.1 - 5 452 Chr. 20 - 49278670 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU536433.1 - 4 423 Chr. 20 - 49278640 - 49289256 [+] on Build GRCh38
          GenBankmRNA EL947487.1 - 5 496 Chr. 20 - 49278667 - 49289243 [+] on Build GRCh38
          GenBankmRNA BP398633.1 - 5 460 Chr. 20 - 49278662 - 49289256 [+] on Build GRCh38
          GenBankmRNA BG530296.1 - 5 482 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA EL950104.1 - 4 398 Chr. 20 - 49278662 - 49289249 [+] on Build GRCh38
          GenBankmRNA BU170823.1 - 5 465 Chr. 20 - 49278657 - 49289256 [+] on Build GRCh38
          GenBankmRNA BU537754.1 - 4 421 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU927979.1 - 3 293 Chr. 20 - 49279156 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU943714.1 - 421 Chr. - - [-] on Build
          GenBankmRNA CN295716.1 - 5 462 Chr. 20 - 49278660 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA335799.1 - 3 241 Chr. 20 - 49280544 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD173681.1 - 3 354 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295717.1 - 4 354 Chr. 20 - 49279155 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU541257.1 - 406 Chr. - - [-] on Build
          GenBankmRNA BP394540.1 - 4 355 Chr. 20 - 49279154 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN353468.1 - 4 411 Chr. 20 - 49278651 - 49289257 [+] on Build GRCh38
          GenBankmRNA BU163963.1 - 467 Chr. - - [-] on Build
          GenBankmRNA BG774276.1 - 5 452 Chr. 20 - 49278669 - 49289259 [+] on Build GRCh38
          GenBankmRNA BU953221.1 - 4 421 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AA313204.1 - 5 453 Chr. 20 - 49278669 - 49289227 [+] on Build GRCh38
          GenBankmRNA CA337851.1 - 4 419 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM738735.1 - 4 423 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA T33971.1 - 213 Chr. - - [-] on Build
          GenBankmRNA CR746822.1 - 4 359 Chr. 20 - 49279150 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD176105.1 - 5 498 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CB124746.1 - 5 480 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY074449.1 - 4 417 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM829996.1 - 5 480 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA BE259966.1 - 440 Chr. - - [-] on Build
          GenBankmRNA AA149846.1 - 5 466 Chr. 20 - 49278655 - 49289250 [+] on Build GRCh38
          GenBankmRNA BF978934.1 - 418 Chr. - - [-] on Build
          GenBankmRNA CN353460.1 - 4 404 Chr. 20 - 49278656 - 49289260 [+] on Build GRCh38
          GenBankmRNA BI226376.1 - 454 Chr. - - [-] on Build
          GenBankmRNA AA315192.1 - 4 421 Chr. 20 - 49278661 - 49289256 [+] on Build GRCh38
          GenBankmRNA BM714564.1 - 5 444 Chr. 20 - 49278678 - 49289251 [+] on Build GRCh38
          GenBankmRNA BU535035.1 - 4 421 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA C14918.1 - 397 Chr. 15 - 34142268 - 34142342 [-] on Build GRCh38
          GenBankmRNA BU537518.1 - 3 335 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG290892.1 - 4 387 Chr. 20 - 49278672 - 49289260 [+] on Build GRCh38
          GenBankmRNA BE269507.1 - 434 Chr. - - [-] on Build
          GenBankmRNA BP395379.1 - 5 437 Chr. 20 - 49278685 - 49289259 [+] on Build GRCh38
          GenBankmRNA BU955244.1 - 3 319 Chr. 20 - 49279130 - 49289245 [+] on Build GRCh38
          GenBankmRNA CN295713.1 - 3 313 Chr. 20 - 49278669 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM309872.1 - 4 389 Chr. 20 - 49278678 - 49289243 [+] on Build GRCh38
          GenBankmRNA BM713580.1 - 4 376 Chr. 20 - 49278684 - 49289260 [+] on Build GRCh38
          GenBankmRNA CF123477.1 - 4 456 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM876538.1 - 5 450 Chr. 20 - 49278679 - 49289251 [+] on Build GRCh38
          GenBankmRNA CN353467.1 - 4 412 Chr. 20 - 49278648 - 49289249 [+] on Build GRCh38
          GenBankmRNA BQ276539.1 - 484 Chr. - - [-] on Build
          GenBankmRNA BG108581.1 - 3 292 Chr. 20 - 49278707 - 49289260 [+] on Build GRCh38
          GenBankmRNA BQ678601.1 - 5 445 Chr. 20 - 49278677 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP398003.1 - 3 296 Chr. 20 - 49279151 - 49289256 [+] on Build GRCh38
          GenBankmRNA CD175919.1 - 3 356 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AV706790.2 - 466 Chr. - - [-] on Build
          GenBankmRNA BM785489.1 - 5 480 Chr. 20 - 49278642 - 49289251 [+] on Build GRCh38
          GenBankmRNA CA406197.1 - 4 385 Chr. 20 - 49278673 - 49289260 [+] on Build GRCh38
          GenBankmRNA BE620459.1 - 5 434 Chr. 20 - 49278689 - 49289260 [+] on Build GRCh38
          GenBankmRNA AV755423.1 - 413 Chr. - - [-] on Build
          GenBankmRNA DW451942.1 - 3 349 Chr. 20 - 49278642 - 49289219 [+] on Build GRCh38
          GenBankmRNA AV703438.1 - 5 466 Chr. 20 - 49278656 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM739693.1 - 5 431 Chr. 20 - 49278690 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM465236.1 - 3 336 Chr. 20 - 49279112 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP396956.1 - 3 254 Chr. 20 - 49280531 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM746639.1 - 5 504 Chr. 20 - 49278641 - 49289209 [+] on Build GRCh38
          GenBankmRNA CD676408.1 - 4 379 Chr. 20 - 49279130 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD299123.1 - 421 Chr. - - [-] on Build
          GenBankmRNA BG170931.1 - 5 463 Chr. 20 - 49278658 - 49289260 [+] on Build GRCh38
          GenBankmRNA CB138604.1 - 5 479 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295714.1 - 2 272 Chr. 20 - 49279104 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD048781.1 - 6 767 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM787355.1 - 4 419 Chr. 20 - 49278643 - 49289251 [+] on Build GRCh38
          GenBankmRNA W94800.1 - 456 Chr. - - [-] on Build
          GenBankmRNA CN353466.1 - 4 388 Chr. 20 - 49278672 - 49289256 [+] on Build GRCh38
          GenBankmRNA HY094146.1 - 4 421 Chr. 20 - 49278643 - 49289247 [+] on Build GRCh38
          GenBankmRNA AV744522.1 - 4 459 Chr. 20 - 49278642 - 49289232 [+] on Build GRCh38
          GenBankmRNA BE897331.1 - 4 399 Chr. 20 - 49278661 - 49289257 [+] on Build GRCh38
          GenBankmRNA BY994496.1 - 4 390 Chr. 20 - 49278670 - 49289260 [+] on Build GRCh38
          GenBankmRNA BI855943.1 - 4 386 Chr. 20 - 49278674 - 49289260 [+] on Build GRCh38
          GenBankmRNA N41712.1 - 5 475 Chr. 20 - 49278669 - 49289250 [+] on Build GRCh38
          GenBankmRNA W76265.1 - 400 Chr. - - [-] on Build
          GenBankmRNA BX463424.2 - 5 447 Chr. 20 - 49278675 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD103693.1 - 4 426 Chr. 20 - 49278640 - 49289256 [+] on Build GRCh38
          GenBankmRNA N47913.1 - 4 386 Chr. 20 - 49279123 - 49289260 [+] on Build GRCh38
          GenBankmRNA BE888655.1 - 4 383 Chr. 20 - 49278677 - 49289252 [+] on Build GRCh38
          GenBankmRNA CB140008.1 - 5 479 Chr. 20 - 49278643 - 49289236 [+] on Build GRCh38
          GenBankmRNA BM768265.1 - 5 485 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA AV756131.1 - 337 Chr. - - [-] on Build
          GenBankmRNA BE268167.1 - 5 431 Chr. 20 - 49278688 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM476605.1 - 466 Chr. - - [-] on Build
          GenBankmRNA W80527.1 - 343 Chr. - - [-] on Build
          GenBankmRNA BM799081.1 - 3 320 Chr. 20 - 49278654 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA336246.1 - 4 422 Chr. 20 - 49278638 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW573582.1 - 3 248 Chr. 20 - 49280537 - 49289260 [+] on Build GRCh38
          GenBankmRNA CK005224.1 - 4 432 Chr. 20 - 49278628 - 49289256 [+] on Build GRCh38
          GenBankmRNA AA187121.1 - 4 391 Chr. 20 - 49278668 - 49289243 [+] on Build GRCh38
          GenBankmRNA H48683.1 - 4 389 Chr. 20 - 49278672 - 49289254 [+] on Build GRCh38
          GenBankmRNA BG112453.1 - 5 466 Chr. 20 - 49278659 - 49289242 [+] on Build GRCh38
          GenBankmRNA BE539018.1 - 3 350 Chr. 20 - 49279104 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP397712.1 - 441 Chr. - - [-] on Build
          GenBankmRNA BQ267294.1 - 276 Chr. - - [-] on Build
          GenBankmRNA DW464895.1 - 3 309 Chr. 20 - 49280492 - 49289248 [+] on Build GRCh38
          GenBankmRNA BU600546.1 - 4 422 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM746749.1 - 5 480 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA BU076692.1 - 5 532 Chr. 20 - 49278271 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY089030.1 - 5 480 Chr. 20 - 49278643 - 49289226 [+] on Build GRCh38
          GenBankmRNA BG034878.1 - 5 448 Chr. 20 - 49278673 - 49289260 [+] on Build GRCh38
          GenBankmRNA CB138600.1 - 5 479 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD247325.1 - 499 Chr. - - [-] on Build
          GenBankmRNA CD249562.1 - 3 350 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW954248.1 - 404 Chr. - - [-] on Build
          GenBankmRNA BU502763.1 - 466 Chr. - - [-] on Build
          GenBankmRNA CD174156.1 - 5 498 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BE876452.1 - 5 448 Chr. 20 - 49278693 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD247470.1 - 5 497 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY145487.1 - 4 417 Chr. 20 - 49278643 - 49289256 [+] on Build GRCh38
          GenBankmRNA CR744518.1 - 4 391 Chr. 20 - 49278669 - 49289227 [+] on Build GRCh38
          GenBankmRNA CA455555.1 - 3 255 Chr. 20 - 49280530 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG831370.1 - 467 Chr. - - [-] on Build
          GenBankmRNA AJ709616.1 - 4 391 Chr. 20 - 49278669 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295712.1 - 3 324 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG621770.1 - 504 Chr. - - [-] on Build
          GenBankmRNA BP397308.1 - 444 Chr. - - [-] on Build
          GenBankmRNA BP422065.1 - 4 416 Chr. 20 - 49278666 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA455477.1 - 484 Chr. - - [-] on Build
          GenBankmRNA AA306121.1 - 5 433 Chr. 20 - 49278689 - 49289250 [+] on Build GRCh38
          GenBankmRNA HY137317.1 - 4 420 Chr. 20 - 49278640 - 49289232 [+] on Build GRCh38
          GenBankmRNA CD173247.1 - 3 348 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA395902.1 - 4 407 Chr. 20 - 49278653 - 49289256 [+] on Build GRCh38
          GenBankmRNA CD174147.1 - 3 351 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY150364.1 - 3 331 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA AA173594.1 - 3 252 Chr. 20 - 49280533 - 49289257 [+] on Build GRCh38
          GenBankmRNA CA337728.1 - 4 462 Chr. 20 - 49278628 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU565045.1 - 4 421 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AA400349.1 - 4 406 Chr. 20 - 49278661 - 49289230 [+] on Build GRCh38
          GenBankmRNA BU957909.1 - 5 483 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA BI870232.1 - 5 478 Chr. 20 - 49278662 - 49289260 [+] on Build GRCh38
          GenBankmRNA D56984.1 - 3 306 Chr. 20 - 49280485 - 49289256 [+] on Build GRCh38
          GenBankmRNA CD176085.1 - 4 439 Chr. 20 - 49278640 - 49289260 [+] on Build GRCh38
          GenBankmRNA BY797059.2 - 5 464 Chr. 20 - 49278658 - 49289257 [+] on Build GRCh38
          GenBankmRNA CB148257.1 - 4 418 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA AV705208.1 - 4 401 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP395366.1 - 3 282 Chr. 20 - 49280503 - 49289252 [+] on Build GRCh38
          GenBankmRNA BU564699.1 - 4 421 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA CN295718.1 - 4 424 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA F29827.1 - 3 306 Chr. 20 - 49278675 - 49289209 [+] on Build GRCh38
          GenBankmRNA BG029385.1 - 409 Chr. - - [-] on Build
          GenBankmRNA AV735685.1 - 4 460 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU954247.1 - 4 422 Chr. 20 - 49278639 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU587925.1 - 2 187 Chr. 20 - 49279175 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW969795.1 - 517 Chr. - - [-] on Build
          GenBankmRNA CN353463.1 - 5 471 Chr. 20 - 49278658 - 49289231 [+] on Build GRCh38
          GenBankmRNA BU963558.1 - 4 421 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD174462.1 - 5 502 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD174831.1 - 5 504 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM840527.1 - 5 481 Chr. 20 - 49278643 - 49289227 [+] on Build GRCh38
          GenBankmRNA BI199049.1 - 5 461 Chr. 20 - 49278662 - 49289260 [+] on Build GRCh38
          GenBankmRNA BX464754.2 - 5 447 Chr. 20 - 49278675 - 49289259 [+] on Build GRCh38
          GenBankmRNA CD678954.1 - 4 399 Chr. 20 - 49278661 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU594883.1 - 5 483 Chr. 20 - 49278642 - 49289259 [+] on Build GRCh38
          GenBankmRNA W73489.1 - 452 Chr. - - [-] on Build
          GenBankmRNA C15238.1 - 396 Chr. 16 - 77005189 - 77005267 [-] on Build GRCh38
          GenBankmRNA AJ715322.1 - 4 391 Chr. 20 - 49278691 - 49289227 [+] on Build GRCh38
          GenBankmRNA AA429766.1 - 5 454 Chr. 20 - 49278667 - 49289211 [+] on Build GRCh38
          GenBankmRNA BU589174.1 - 3 335 Chr. 20 - 49278642 - 49289265 [+] on Build GRCh38
          GenBankmRNA BM745349.1 - 5 483 Chr. 20 - 49278640 - 49289219 [+] on Build GRCh38
          GenBankmRNA BI834636.1 - 5 469 Chr. 20 - 49278653 - 49289260 [+] on Build GRCh38
          GenBankmRNA AA102834.1 - 4 401 Chr. 20 - 49278662 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN353462.1 - 4 416 Chr. 20 - 49278646 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY067680.1 - 4 423 Chr. 20 - 49278640 - 49289236 [+] on Build GRCh38
          GenBankmRNA W57650.1 - 406 Chr. - - [-] on Build
          GenBankmRNA HY111069.1 - 5 479 Chr. 20 - 49278643 - 49289256 [+] on Build GRCh38
          GenBankmRNA AW965842.1 - 452 Chr. - - [-] on Build
          GenBankmRNA BM841244.1 - 5 482 Chr. 20 - 49278643 - 49289239 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gbgwf:80/100.66.79.18:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0