Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC101927136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110840.1 | 4 | 720 | |||
GenBank mRNA | AK125128.1 | - | 4 | 930 | ||
AK126334.1 | - | 5 | 795 | |||
BC050580.1 | - | 4 | 696 |