Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022317.3 | NP_071712.3 | 11 | 1157 | ||
GenBank mRNA | AF305211.1 | - | 11 | 1090 | ||
AK052784.1 | - | 11 | 1158 | |||
AK145077.1 | - | 11 | 1158 | |||
BC010472.1 | - | 11 | 1145 | 3586005 | ||
BC013783.1 | - | 11 | 1148 | 3600786 |