Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054097.3 | NP_473438.1 | 2, 3 | 378 | ||
GenBank mRNA | AB054591.1 | - | 2, 3 | 321 | ||
AK140079.1 | - | 2, 3 | 342 | |||
AK182167.1 | - | 1, 2 | 71 | |||
BC021383.1 | - | 2, 3 | 311 | 5042933 |