Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PARK2 SIRNA Assays ›
          Gene Symbol:
          PARK2
          Gene Name:
          parkin RBR E3 ubiquitin protein ligase
          Gene Aliases:
          AR-JP, LPRS2, PDJ, PRKN
          Chromosome Location:
          Chr.6: 161347417 - 162727802 on Build GRCh38
          UniGene ID:
          Hs.132954
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10044

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PARK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004562.2 NP_004553.2 7 922
          NM_013987.2 NP_054642.2 6 838
          NM_013988.2 NP_054643.2 4 475
          XM_011535863.1 XP_011534165.1 7 919
          XM_017010908.1 XP_016866397.1 7 1836
          XM_017010909.1 XP_016866398.1 6 716
          GenBank mRNA AB009973.1 - 7 889
          AB245403.1 - 8 880
          AF381282.1 - 5 547
          AF381283.1 - 5 624
          AF381284.1 - 5 666
          AK292590.1 - 7 883
          AK294684.1 - 2 128
          BC022014.2 - 7 886 4824892
          EF375726.1 - 6 788
          GU345837.1 - 6 641
          GU345838.1 - 5 683
          GU345839.1 - 6 721
          GU345840.1 - 6 805
          GU357501.1 - 4 278
          GU357502.1 - 3 278
          GU361466.1 - 5 683
          GU361467.1 - 5 721
          GU361468.1 - 5 564
          GU361469.1 - 7 902
          GU361470.1 - 7 903
          KC357595.1 - 9 1000
          KC774171.1 - 5 655
          KJ534911.1 - 6 788
          KJ534912.1 - 6 788
          KU178240.1 - 6 788
          KU178241.1 - 6 788
          KU178242.1 - 6 785

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein polyubiquitination
          mitochondrial fission
          mitophagy
          negative regulation of protein phosphorylation
          startle response
          transcription, DNA-templated
          protein monoubiquitination
          response to oxidative stress
          mitochondrion organization
          central nervous system development
          learning
          adult locomotory behavior
          proteasomal protein catabolic process
          regulation of autophagy
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of mitochondrial fusion
          negative regulation of mitochondrial fusion
          regulation of mitochondrion organization
          regulation of glucose metabolic process
          free ubiquitin chain polymerization
          regulation of dopamine secretion
          macroautophagy
          protein ubiquitination
          regulation of protein ubiquitination
          protein destabilization
          negative regulation of actin filament bundle assembly
          regulation of lipid transport
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of glucokinase activity
          cellular response to unfolded protein
          response to endoplasmic reticulum stress
          synaptic transmission, glutamatergic
          protein K29-linked ubiquitination
          ERAD pathway
          regulation of dopamine metabolic process
          norepinephrine metabolic process
          dopamine metabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          cellular protein catabolic process
          cellular protein metabolic process
          protein K27-linked ubiquitination
          negative regulation by host of viral genome replication
          positive regulation of protein catabolic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of JNK cascade
          negative regulation of insulin secretion
          protein stabilization
          positive regulation of neurotransmitter uptake
          dopamine uptake involved in synaptic transmission
          protein autoubiquitination
          regulation of mitochondrial membrane potential
          zinc ion homeostasis
          negative regulation of cell death
          regulation of canonical Wnt signaling pathway
          parkin-mediated mitophagy in response to mitochondrial depolarization
          neuron cellular homeostasis
          protein K63-linked ubiquitination
          protein localization to mitochondrion
          aggresome assembly
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          cellular response to manganese ion
          protein K6-linked ubiquitination
          negative regulation of canonical Wnt signaling pathway
          positive regulation of mitochondrial fission
          negative regulation of release of cytochrome c from mitochondria
          cellular response to toxic substance
          mitophagy in response to mitochondrial depolarization
          regulation of cellular response to oxidative stress
          negative regulation of neuron death
          positive regulation of proteasomal protein catabolic process
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          negative regulation of primary amine oxidase activity
          positive regulation of protein linear polyubiquitination
          regulation of synaptic vesicle transport
          negative regulation of oxidative stress-induced cell death
          regulation of protein targeting to mitochondrion
          positive regulation of tumor necrosis factor-mediated signaling pathway
          cellular response to dopamine
          negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of dendrite extension
          negative regulation of spontaneous neurotransmitter secretion
          regulation of reactive oxygen species metabolic process
          negative regulation of reactive oxygen species metabolic process
          transcription regulatory region sequence-specific DNA binding
          G-protein coupled receptor binding
          transcription factor activity, sequence-specific DNA binding
          actin binding
          ubiquitin-protein transferase activity
          protein binding
          beta-catenin binding
          zinc ion binding
          tubulin binding
          ligase activity
          SH3 domain binding
          enzyme binding
          kinase binding
          protein kinase binding
          PDZ domain binding
          Hsp70 protein binding
          heat shock protein binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          identical protein binding
          histone deacetylase binding
          ubiquitin binding
          phospholipase binding
          chaperone binding
          ubiquitin protein ligase activity
          cullin family protein binding
          ubiquitin protein ligase activity involved in ERAD pathway
          ubiquitin-specific protease binding
          F-box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zshvn:80/100.66.77.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0