Search
Search

Igsf8| Gene Symbol: Igsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080419.1 | NP_536344.1 | 3 | 876 | ||
| GenBank mRNA | AF411055.1 | - | 3 | 747 | ||
| AF439263.1 | - | 3 | 859 | |||
| AK140254.1 | - | 3 | 889 | |||
| AK146104.1 | - | 3 | 889 | |||
| AK146221.1 | - | 3 | 895 | |||
| AK150239.1 | - | 3 | 895 | |||
| AK158522.1 | - | 3 | 897 | |||
| AK168363.1 | - | 3 | 895 | |||
| AK178388.1 | - | 1 | 74 | |||
| AK203486.1 | - | 1 | 73 | |||
| AK213984.1 | - | 1 | 324 | |||
| BC026464.1 | - | 2 | 682 | |||
| BC038068.1 | - | 1 | 133 | |||
| BC048387.1 | - | 3 | 876 | 6311441 | ||
| BC060087.1 | - | 1504 | ||||