Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130888.1 | NP_570958.1 | 10 | 862 | ||
GenBank mRNA | AB178901.1 | - | 11 | 1001 | ||
AJ305318.1 | - | 7 | 531 | |||
AY260550.1 | - | 10 | 862 |