Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005388.4 | NP_005379.3 | 4 | 744 | ||
XM_017014785.1 | XP_016870274.1 | 3 | 520 | |||
GenBank mRNA | AF031463.1 | - | 4 | 721 | ||
AK024980.1 | - | 1 | 20 | |||
AK313429.1 | - | 4 | 744 | |||
AL117602.1 | - | 4 | 677 | |||
BC017227.2 | - | 4 | 619 | 3948368 |