Search
Search

Bspry| Gene Symbol: Bspry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138653.1 | NP_619594.1 | 3 | 628 | ||
| GenBank mRNA | AJ276690.1 | - | 3 | 628 | ||
| AK035588.1 | - | 3 | 634 | |||
| AK160930.1 | - | 3 | 634 | |||
| BC003992.1 | - | 1 | 171 | |||
| BC139079.1 | - | 3 | 644 | 8862101 | ||
| BC139080.1 | - | 3 | 644 | 8862102 | ||
| BC145535.1 | - | 3 | 644 | 9054098 | ||