Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IDO2 SIRNA Assays ›
          Gene Symbol:
          Ido2
          Gene Name:
          indoleamine 2,3-dioxygenase 2
          Gene Aliases:
          AI265623, C230043N17Rik, Ido-2, Indol1
          Chromosome Location:
          Chr.8: 24531892 - 24576404 on Build GRCm38
          UniGene ID:
          Mm.219580
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s102024

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ido2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_145949.2 NP_666061.3 4, 6 526
          GenBank mRNA AK082385.1 - 3, 5 717
          BC026393.1 - 4, 6 526 4195134
          EF137182.1 - 4, 6 526
          EU440733.1 - 3, 5 318

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          tryptophan catabolic process to kynurenine
          oxidation-reduction process
          tryptophan 2,3-dioxygenase activity
          oxidoreductase activity
          2,3-dihydroxy DDT 1,2-dioxygenase activity
          phenanthrene dioxygenase activity
          2,2',3-trihydroxybiphenyl dioxygenase activity
          1,2-dihydroxyfluorene 1,1-alpha-dioxygenase activity
          5,6-dihydroxy-3-methyl-2-oxo-1,2-dihydroquinoline dioxygenase activity
          1,1-dichloro-2-(dihydroxy-4-chlorophenyl)-(4-chlorophenyl)ethene 1,2-dioxygenase activity
          protocatechuate 3,4-dioxygenase type II activity
          2'-aminobiphenyl-2,3-diol 1,2-dioxygenase activity
          3,4-dihydroxyfluorene 4,4-alpha-dioxygenase activity
          2,3-dihydroxy-ethylbenzene 1,2-dioxygenase activity
          carbazole 1,9a-dioxygenase activity
          dihydroxydibenzothiophene dioxygenase activity
          1,2-dihydroxynaphthalene-6-sulfonate 1,8a-dioxygenase activity
          styrene dioxygenase activity
          3,4-dihydroxyphenanthrene dioxygenase activity
          hydroquinone 1,2-dioxygenase activity
          p-cumate 2,3-dioxygenase activity
          2,3-dihydroxy-p-cumate dioxygenase activity
          3,5-dichlorocatechol 1,2-dioxygenase activity
          2-aminophenol 1,6-dioxygenase activity
          2,6-dichloro-p-hydroquinone 1,2-dioxygenase activity
          chlorocatechol 1,2-dioxygenase activity
          catechol dioxygenase activity
          dihydroxyfluorene dioxygenase activity
          heme binding
          indoleamine 2,3-dioxygenase activity
          5-aminosalicylate dioxygenase activity
          3-hydroxy-2-naphthoate 2,3-dioxygenase activity
          benzo(a)pyrene 11,12-dioxygenase activity
          benzo(a)pyrene 4,5-dioxygenase activity
          4,5-dihydroxybenzo(a)pyrene dioxygenase activity
          benzo(a)pyrene 9,10-dioxygenase activity
          9,10-dihydroxybenzo(a)pyrene dioxygenase activity
          benzo(a)pyrene 7,8-dioxygenase activity
          7,8-dihydroxy benzo(a)pyrene dioxygenase activity
          1,2-dihydroxy-5,6,7,8-tetrahydronaphthalene extradiol dioxygenase activity
          2-mercaptobenzothiazole dioxygenase activity
          pyridine-3,4-diol dioxygenase activity
          pyrene dioxygenase activity
          4,5-dihydroxypyrene dioxygenase activity
          phenanthrene-4-carboxylate dioxygenase activity
          tetrachlorobenzene dioxygenase activity
          4,6-dichloro-3-methylcatechol 1,2-dioxygenase activity
          2,3-dihydroxydiphenyl ether dioxygenase activity
          diphenyl ether 1,2-dioxygenase activity
          arachidonate 8(S)-lipoxygenase activity
          metal ion binding
          4-hydroxycatechol 1,2-dioxygenase activity
          dioxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2lcrp:80/100.66.72.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0