Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Thsd7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172485.3 | NP_766073.2 | 7 | 1988 | ||
GenBank mRNA | AK051714.1 | - | 7 | 1830 | ||
AK083798.1 | - | 7 | 1816 | |||
BC063250.1 | - | 7 | 1818 | 6410178 |