Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wwc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170779.1 | NP_740749.1 | 22 | 3149 | ||
GenBank mRNA | AK220259.1 | - | 3520 | |||
BC006733.1 | - | 19 | 2738 | |||
BC017638.1 | - | 15 | 2275 | |||
BC037006.1 | - | 14 | 1997 | 5116674 | ||
DQ256090.1 | - | 22 | 3149 |