Search
Search

Cspp1| Gene Symbol: Cspp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026493.3 | NP_080769.3 | 29 | 3442 | ||
| GenBank mRNA | BC055915.1 | - | 11 | 1146 | ||
| BC151042.1 | - | 29 | 3415 | 9087955 | ||
| BM949181.1 | - | 3 | 217 | |||
| FJ008676.1 | - | 29 | 3140 | |||