Search
Search

Snx13| Gene Symbol: Snx13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014973.2 | NP_001014973.2 | 18 | 1943 | ||
| GenBank mRNA | AK122352.1 | - | 7 | 592 | ||
| AK133353.1 | - | 15 | 1483 | |||
| AK154425.1 | - | 18 | 1945 | |||
| AK164161.1 | - | 18 | 1868 | |||
| AK166180.1 | - | 17 | 1836 | |||
| BC056394.1 | - | 18 | 1915 | 6852346 | ||
| BC067201.1 | - | 18 | 1932 | 6825168 | ||