Search
Search

Tmem55b| Gene Symbol: Tmem55b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033271.4 | NP_001028443.1 | 2 | 394 | ||
| GenBank mRNA | AK144251.1 | - | 2 | 394 | ||
| AK152640.1 | - | 2 | 380 | |||
| AK152658.1 | - | 2 | 380 | |||
| AK159641.1 | - | 2 | 398 | |||
| AK169026.1 | - | 2 | 376 | |||
| BC039161.1 | - | 2 | 337 | |||
| BC049945.1 | - | 2 | 357 | |||
| BC058171.1 | - | 2 | 343 | |||
| BC106163.1 | - | 2 | 373 | 4913757 | ||