Search
Search

Alg12| Gene Symbol: Alg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142357.1 | NP_001135829.1 | 9 | 1285 | ||
| NM_145477.2 | NP_663452.1 | 7 | 982 | |||
| GenBank mRNA | AJ429133.1 | - | 9 | 1245 | ||
| AK142322.1 | - | 9 | 1286 | |||
| BC021379.1 | - | 7 | 955 | 5044319 | ||