Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab40c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139154.2 | NP_631893.1 | 5 | 437 | ||
GenBank mRNA | AB232639.1 | - | 4 | 331 | ||
AF422144.1 | - | 4 | 390 | |||
AK133766.1 | - | 4 | 213 | |||
BC027008.1 | - | 5 | 425 | 5364768 | ||
BC037461.1 | - | 6 | 869 |