Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRP4 SIRNA Assays ›
          Gene Symbol:
          Lrp4
          Gene Name:
          low density lipoprotein receptor-related protein 4
          Gene Aliases:
          6430526J12Rik, D230026E03, Megf7, mdig
          Chromosome Location:
          Chr.2: 91457470 - 91513901 on Build GRCm38
          UniGene ID:
          Mm.275149
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s105882

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lrp4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_172668.3 NP_766256.3 38 5683
          GenBank mRNA AF247637.1 - 38 5513
          AK129224.1 - 24 3472
          BC132240.1 - 38 5606 40130517

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031322, AB073317, AB011533.

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          regulation of protein phosphorylation
          hair follicle development
          endocytosis
          multicellular organism development
          protein localization
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          Wnt signaling pathway
          cell differentiation
          negative regulation of Wnt signaling pathway
          negative regulation of ossification
          embryonic limb morphogenesis
          odontogenesis of dentin-containing tooth
          embryonic digit morphogenesis
          receptor clustering
          Wnt signaling pathway involved in dorsal/ventral axis specification
          dendrite morphogenesis
          anatomical structure development
          negative regulation of axonogenesis
          synapse organization
          synaptic growth at neuromuscular junction
          protein heterotetramerization
          canonical Wnt signaling pathway
          limb development
          skeletal muscle acetylcholine-gated channel clustering
          negative regulation of canonical Wnt signaling pathway
          postsynaptic membrane assembly
          presynaptic membrane assembly
          positive regulation of presynaptic membrane organization
          calcium ion binding
          protein binding
          Wnt-protein binding
          receptor tyrosine kinase binding
          apolipoprotein binding
          protein homodimerization activity
          Wnt-activated receptor activity
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hgr46:80/100.66.79.101:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0