Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045523.1 | NP_001038988.1 | 6 | 2249 | ||
GenBank mRNA | AK129243.1 | - | 6 | 2407 | ||
AK152597.1 | - | 6 | 2188 | |||
BC047433.1 | - | 5 | 998 | |||
BC100358.1 | - | 6 | 2411 | 30848279 | ||
BC141046.1 | - | 5 | 2112 | 9055658 | ||
BC145233.1 | - | 5 | 2103 | 9053785 |