Search
Search

Galnt12| Gene Symbol: Galnt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172693.3 | NP_766281.1 | 6 | 1115 | ||
| GenBank mRNA | AB175680.1 | - | 6 | 1095 | ||
| AK033638.1 | - | 6 | 1116 | |||
| AK042133.1 | - | 6 | 1148 | |||
| AK043977.1 | - | 5 | 976 | |||
| AK082014.1 | - | 6 | 1161 | |||
| AK156751.1 | - | 6 | 1145 | |||
| BC024475.1 | - | 5 | 947 | |||
| BC025639.1 | - | 5 | 837 | |||
| BC056425.1 | - | 6 | 1126 | 5716909 | ||