Search
Search

Pdik1l| Gene Symbol: Pdik1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163794.1 | NP_001157266.1 | 3 | 1022 | ||
| NM_146156.3 | NP_666268.1 | 3 | 1149 | |||
| GenBank mRNA | AK139413.1 | - | 3 | 1019 | ||
| AK141566.1 | - | 3 | 1012 | |||
| AK141823.1 | - | 3 | 1023 | |||
| AK157013.1 | - | 3 | 995 | |||
| AK164975.1 | - | 3 | 1201 | |||
| BC027088.1 | - | 3 | 996 | 5358000 | ||
| BC058514.1 | - | 3 | 1135 | 6821637 | ||