Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040631.2 | NP_001035721.1 | 3 | 648 | ||
NM_001040632.2 | NP_001035722.1 | 3 | 739 | |||
NM_001190313.1 | NP_001177242.1 | 14 | 3646 | |||
NM_001190319.1 | NP_001177248.1 | 3 | 723 | |||
NM_145613.4 | NP_663588.2 | 14 | 3664 | |||
GenBank mRNA | AF469650.1 | - | 14 | 3592 | ||
AK053629.1 | - | 14 | 3648 | |||
BC023068.1 | - | 3 | 648 | 5355789 | ||
BC025174.1 | - | 3 | 607 | 3978387 | ||
BE861753.1 | - | 1 | 108 | |||
DQ002398.1 | - | 3 | 647 |
Gene Symbol: Mfrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190314.1 | NP_001177243.1 | 14 | 3646 | ||
NM_147126.3 | NP_667337.1 | 14 | 3664 |