Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000446.5 | NP_000437.3 | 3, 4 | 307 | ||
GenBank mRNA | AK314027.1 | - | 3, 4 | 233 | ||
BC074719.2 | - | 3, 4 | 239 | 30915169 | ||
BJ993711.1 | - | 3, 4 | 307 | |||
CB122285.1 | - | 3, 4 | 267 | |||
D84371.1 | - | 3, 4 | 264 | |||
M63012.1 | - | 3, 4 | 219 | |||
M63013.1 | - | 3, 4 | 158 | |||
M63014.1 | - | 2, 3 | 124 | |||
S64615.1 | - | 3, 4 | 219 | |||
U53784.1 | - | 3, 4 | 264 | |||
Z70723.1 | - | 3, 4 | 264 |