Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOX1 SIRNA Assays ›
          Gene Symbol:
          Nox1
          Gene Name:
          NADPH oxidase 1
          Gene Aliases:
          GP91-2, MOX1, NOH-1, NOH1, NOX1a, NOX1alpha, Nox-1
          Chromosome Location:
          Chr.X: 134086012 - 134137711 on Build GRCm38
          UniGene ID:
          Mm.233865
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s108444

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nox1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_172203.2 NP_757340.1 2 265
          GenBank mRNA AB206383.1 - 2 265
          AB206384.1 - 4 517
          AB206385.1 - 674
          AF539799.1 - 2 119
          AK136432.1 - 2 180

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053683, AF152963.

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          angiogenesis
          regulation of systemic arterial blood pressure by renin-angiotensin
          superoxide metabolic process
          transport
          signal transduction
          positive regulation of cell proliferation
          proton transport
          cell migration
          extracellular matrix organization
          regulation of ion transmembrane transport
          superoxide anion generation
          hydrogen peroxide metabolic process
          positive regulation of MAPK cascade
          positive regulation of integrin biosynthetic process
          positive regulation of JNK cascade
          positive regulation of smooth muscle cell proliferation
          intracellular pH elevation
          oxidation-reduction process
          cellular response to hyperoxia
          oxygen metabolic process
          cation transmembrane transport
          positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          cellular stress response to acidic pH
          positive regulation of reactive oxygen species metabolic process
          N-ethylmaleimide reductase activity
          superoxide-generating NADPH oxidase activity
          oxidoreductase activity
          voltage-gated proton channel activity
          reduced coenzyme F420 dehydrogenase activity
          sulfur oxygenase reductase activity
          malolactic enzyme activity
          NADPH:sulfur oxidoreductase activity
          metal ion binding
          Rac GTPase binding
          epoxyqueuosine reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-66c8j:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0