Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fancb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146081.1 | NP_001139553.1 | 3 | 1270 | ||
NM_175027.4 | NP_778192.3 | 3 | 1270 | |||
GenBank mRNA | AK132251.1 | - | 3 | 1270 | ||
AK169374.1 | - | 3 | 1158 | |||
BC083151.1 | - | 3 | 1163 | 30460698 |