Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FN3KRP SIRNA Assays ›
Gene Symbol:
Fn3krp
Gene Name:
fructosamine 3 kinase related protein
Gene Aliases:
FN3K-RP
Chromosome Location:
Chr.11: 121421373 - 121431288 on Build GRCm38
UniGene ID:
Mm.117394
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s108711
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Fn3krp
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181420.3
NP_852085.2
4
509
GenBank mRNA
AY360466.1
-
4
467
BC032265.1
-
4
455
4015688
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107077, BC166839.
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
phosphorylation
molecular_function
phosphofructokinase activity
phosphorylase kinase regulator activity
cobinamide kinase activity
phytol kinase activity
kinase activity
phosphatidylinositol phosphate kinase activity
cyclin-dependent protein serine/threonine kinase regulator activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phenol kinase activity
cyclin-dependent protein kinase activating kinase regulator activity
inositol tetrakisphosphate 2-kinase activity
heptose 7-phosphate kinase activity
aminoglycoside phosphotransferase activity
phosphatidylinositol 3-kinase activity
eukaryotic elongation factor-2 kinase regulator activity
eukaryotic elongation factor-2 kinase activator activity
LPPG:FO 2-phospho-L-lactate transferase activity
cytidine kinase activity
glycerate 2-kinase activity
(S)-lactate 2-kinase activity
phosphoserine:homoserine phosphotransferase activity
L-seryl-tRNA(Sec) kinase activity
phosphocholine transferase activity
polynucleotide 5'-hydroxyl-kinase activity
ATP-dependent polynucleotide kinase activity
GTP-dependent polynucleotide kinase activity
farnesol kinase activity
CTP:2-trans,-6-trans-farnesol kinase activity
geraniol kinase activity
geranylgeraniol kinase activity
CTP:geranylgeraniol kinase activity
prenol kinase activity
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity
phosphatidylinositol bisphosphate kinase activity
inositol-3,4,6-trisphosphate 1-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-vs5rc:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0