Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POU6F1 SIRNA Assays ›
          Gene Symbol:
          POU6F1
          Gene Name:
          POU class 6 homeobox 1
          Gene Aliases:
          BRN5, MPOU, TCFB1
          Chromosome Location:
          Chr.12: 51186936 - 51218134 on Build GRCh38
          UniGene ID:
          Hs.555886
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10876

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POU6F1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002702.3 NP_002693.3 4 822
          NR_026893.1 9 1280
          XM_017019507.1 XP_016874996.1 9 2461
          XM_017019508.1 XP_016874997.1 10 2372
          XM_017019509.1 XP_016874998.1 8 1381
          XM_017019510.1 XP_016874999.1 9 1810
          XM_017019511.1 XP_016875000.1 9 1979
          XM_017019512.1 XP_016875001.1 9 1335
          XM_017019513.1 XP_016875002.1 9 2458
          XM_017019514.1 XP_016875003.1 9 2461
          XM_017019515.1 XP_016875004.1 9 1807
          XM_017019516.1 XP_016875005.1 9 2458
          XM_017019517.1 XP_016875006.1 8 2332
          XM_017019518.1 XP_016875007.1 8 2332
          XM_017019519.1 XP_016875008.1 9 2461
          XM_017019520.1 XP_016875009.1 7 1688
          XM_017019521.1 XP_016875010.1 7 1707
          XM_017019522.1 XP_016875011.1 8 1185
          XM_017019523.1 XP_016875012.1 6 892
          XM_017019524.1 XP_016875013.1 8 1289
          XM_017019525.1 XP_016875014.1 9 1277
          XM_017019526.1 XP_016875015.1 8 1185
          XM_017019527.1 XP_016875016.1 8 1204
          XM_017019528.1 XP_016875017.1 9 1710
          XM_017019529.1 XP_016875018.1 8 1596
          XM_017019530.1 XP_016875019.1 7 1501
          GenBank mRNA AK095191.1 - 9 1280
          AL832881.1 - 4 822
          BC051326.1 - 4 625 6275848
          BC074765.2 - 3 365 30915178
          L14482.1 - 4 493
          Z21966.1 - 3 532

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          brain development
          heart development
          muscle organ development
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0