Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPOX SIRNA Assays ›
Gene Symbol:
PPOX
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Chromosome Location:
Chr.1: 161165728 - 161178277 on Build GRCh38
UniGene ID:
Hs.517373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10927
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPOX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000309.3
NP_000300.1
3
381
NM_001122764.1
NP_001116236.1
3
442
XM_005245291.3
XP_005245348.2
3
420
XM_005245295.3
XP_005245352.2
3
365
XM_006711402.2
XP_006711465.2
3
372
XM_006711403.2
XP_006711466.2
3
376
XM_006711404.3
XP_006711467.2
3
309
XM_006711406.2
XP_006711469.2
3
278
XM_011509663.2
XP_011507965.1
3
492
XM_011509664.1
XP_011507966.1
3
278
XM_011509665.2
XP_011507967.1
3
490
XM_011509666.2
XP_011507968.1
3
497
XM_011509667.2
XP_011507969.1
3
433
XM_011509668.2
XP_011507970.1
3
233
XM_011509669.1
XP_011507971.1
3
246
XM_011509670.2
XP_011507972.1
3
495
XM_011509671.1
XP_011507973.1
3
278
XM_011509672.2
XP_011507974.1
3
486
XM_011509673.2
XP_011507975.1
3
493
XM_011509674.2
XP_011507976.1
3
498
XM_011509675.2
XP_011507977.1
3
428
XM_011509676.1
XP_011507978.1
3
437
XM_011509677.1
XP_011507979.1
3
370
XM_011509678.1
XP_011507980.1
3
434
XM_011509679.1
XP_011507981.1
3
242
XM_011509681.1
XP_011507983.1
3
292
XM_011509682.1
XP_011507984.1
3
365
XM_017001559.1
XP_016857048.1
3
489
XM_017001560.1
XP_016857049.1
3
420
XM_017001561.1
XP_016857050.1
3
373
XM_017001562.1
XP_016857051.1
3
347
XM_017001563.1
XP_016857052.1
3
432
XM_017001565.1
XP_016857054.1
3
347
XM_017001566.1
XP_016857055.1
3
424
XM_017001567.1
XP_016857056.1
3
365
XM_017001568.1
XP_016857057.1
3
424
XM_017001569.1
XP_016857058.1
3
292
XM_017001570.1
XP_016857059.1
3
405
XM_017001571.1
XP_016857060.1
3
510
GenBank mRNA
AK094855.1
-
3
296
AK298912.1
-
3
322
AK302298.1
-
3
351
BC002357.2
-
3
362
2821983
BX384814.2
-
3
441
D38537.1
-
3
429
DC329287.1
-
3
453
U26446.1
-
3
343
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00609392_m1
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bfkrn:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0