Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPOX SIRNA Assays ›
          Gene Symbol:
          PPOX
          Gene Name:
          protoporphyrinogen oxidase
          Gene Aliases:
          PPO, V290M, VP
          Chromosome Location:
          Chr.1: 161165728 - 161178277 on Build GRCh38
          UniGene ID:
          Hs.517373
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10928
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPOX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000309.3 NP_000300.1 6 813
          NM_001122764.1 NP_001116236.1 6 874
          XM_005245291.3 XP_005245348.2 6 852
          XM_005245295.3 XP_005245352.2 5 700
          XM_006711402.2 XP_006711465.2 6 819
          XM_006711403.2 XP_006711466.2 6 808
          XM_006711404.3 XP_006711467.2 6 756
          XM_006711406.2 XP_006711469.2 6 729
          XM_011509663.2 XP_011507965.1 6 939
          XM_011509664.1 XP_011507966.1 6 710
          XM_011509665.2 XP_011507967.1 6 937
          XM_011509666.2 XP_011507968.1 6 944
          XM_011509667.2 XP_011507969.1 6 880
          XM_011509668.2 XP_011507970.1 6 680
          XM_011509669.1 XP_011507971.1 6 693
          XM_011509670.2 XP_011507972.1 5 828
          XM_011509671.1 XP_011507973.1 6 710
          XM_011509672.2 XP_011507974.1 5 819
          XM_011509673.2 XP_011507975.1 4 676
          XM_011509674.2 XP_011507976.1 6 945
          XM_011509675.2 XP_011507977.1 5 761
          XM_011509676.1 XP_011507978.1 5 1024
          XM_011509677.1 XP_011507979.1 5 957
          XM_011509678.1 XP_011507980.1 6 917
          XM_011509679.1 XP_011507981.1 6 725
          XM_011509681.1 XP_011507983.1 5 642
          XM_017001559.1 XP_016857048.1 6 936
          XM_017001560.1 XP_016857049.1 6 852
          XM_017001561.1 XP_016857050.1 5 1345
          XM_017001562.1 XP_016857051.1 6 798
          XM_017001563.1 XP_016857052.1 6 896
          XM_017001564.1 XP_016857053.1 2 414
          XM_017001565.1 XP_016857054.1 6 798
          XM_017001566.1 XP_016857055.1 6 907
          XM_017001567.1 XP_016857056.1 5 700
          XM_017001568.1 XP_016857057.1 6 888
          XM_017001569.1 XP_016857058.1 5 642
          GenBank mRNA AK094855.1 - 5 646
          AK302298.1 - 5 684
          BC002357.2 - 6 794 2821983
          D38537.1 - 6 861
          U26446.1 - 6 775

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00609392_m1

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          porphyrin-containing compound biosynthetic process
          protoporphyrinogen IX biosynthetic process
          heme biosynthetic process
          response to drug
          oxidation-reduction process
          oxygen-dependent protoporphyrinogen oxidase activity
          flavin adenine dinucleotide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0