Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc16a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172838.3 | NP_766426.1 | 5 | 864 | ||
GenBank mRNA | AK028284.1 | - | 5 | 862 | ||
AK029076.1 | - | 5 | 864 | |||
BC111902.1 | - | 4 | 817 | 40062303 | ||
BC113803.1 | - | 4 | 817 | 40062304 |