Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172295.4 | NP_758499.3 | 2 | 550 | ||
GenBank mRNA | AK029040.1 | - | 2 | 289 | ||
AK161057.1 | - | 2 | 467 | |||
AK166928.1 | - | 300 | ||||
AK172389.1 | - | 2 | 499 | |||
BC037703.1 | - | 2 | 487 | 4166596 |