Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP2R5C SIRNA Assays ›
Gene Symbol:
PPP2R5C
Gene Name:
protein phosphatase 2 regulatory subunit B'gamma
Gene Aliases:
B56G, PR61G
Chromosome Location:
Chr.14: 101760573 - 101927992 on Build GRCh38
UniGene ID:
Hs.368264
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10988
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
562.00
Your Price
Online offer:
533.65
533.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP2R5C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161725.1
NP_001155197.1
11
1101
NM_001161726.1
NP_001155198.1
11
1173
NM_002719.3
NP_002710.2
9
1052
NM_178586.2
NP_848701.1
9
1052
NM_178587.2
NP_848702.1
9
1052
XM_005267819.1
XP_005267876.1
11
1162
XM_005267820.1
XP_005267877.1
11
1102
XM_005267822.3
XP_005267879.1
11
1162
XM_005267823.1
XP_005267880.1
11
1162
XM_005267824.1
XP_005267881.1
11
1162
XM_005267826.1
XP_005267883.1
9
875
XM_005267827.1
XP_005267884.1
11
1097
XM_011536914.2
XP_011535216.1
10
966
XM_011536915.2
XP_011535217.1
10
1049
XM_011536916.2
XP_011535218.1
10
2048
XM_011536918.1
XP_011535220.1
10
1203
XM_011536919.2
XP_011535221.1
10
1185
XM_011536920.1
XP_011535222.1
10
1079
XM_011536921.2
XP_011535223.1
9
968
XM_017021422.1
XP_016876911.1
10
1118
XM_017021423.1
XP_016876912.1
9
1052
XM_017021424.1
XP_016876913.1
10
1079
GenBank mRNA
AB451341.1
-
9
912
AK131391.1
-
11
1086
AK302470.1
-
11
1001
AK315072.1
-
9
988
BC016183.1
-
9
950
4855950
BX248254.1
-
7
547
D26445.1
-
9
909
L42375.1
-
9
882
U37352.1
-
9
970
Z69030.1
-
9
990
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00604902_m1
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
negative regulation of cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
regulation of protein phosphatase type 2A activity
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
protein phosphatase type 2A regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0