Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006437.3 | NP_006428.2 | 33 | 5008 | ||
XM_011534931.1 | XP_011533233.1 | 32 | 5058 | |||
XM_011534932.2 | XP_011533234.1 | 33 | 5225 | |||
GenBank mRNA | AF057160.1 | - | 33 | 4954 | ||
AF158255.1 | - | 33 | 5008 | |||
D79999.2 | - | 32 | 4907 |