Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKACB SIRNA Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
UniGene ID:
Hs.487325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11068
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKACB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242857.2
NP_001229786.1
9, 10
794
NM_001242858.2
NP_001229787.1
6, 7
737
NM_001242859.2
NP_001229788.1
8, 9
785
NM_001242860.2
NP_001229789.1
9, 10
929
NM_001242861.2
NP_001229790.1
7, 8
821
NM_001242862.2
NP_001229791.1
5, 6
533
NM_001300915.1
NP_001287844.1
9, 10
929
NM_001300916.1
NP_001287845.1
6, 7
799
NM_001300917.1
NP_001287846.1
6, 7
737
NM_002731.3
NP_002722.1
6, 7
895
NM_182948.3
NP_891993.1
6, 7
799
NM_207578.2
NP_997461.1
6, 7
895
XM_005271015.3
XP_005271072.1
8, 9
746
XM_005271016.1
XP_005271073.1
8, 9
871
XM_005271017.3
XP_005271074.1
7, 8
736
XM_005271018.4
XP_005271075.1
7, 8
1056
XM_005271019.1
XP_005271076.1
6, 7
812
XM_005271020.1
XP_005271077.1
6, 7
779
XM_006710758.1
XP_006710821.1
8, 9
885
XM_017001706.1
XP_016857195.1
9, 10
789
XM_017001707.1
XP_016857196.1
9, 10
789
XM_017001708.1
XP_016857197.1
9, 10
890
XM_017001709.1
XP_016857198.1
8, 9
587
XM_017001710.1
XP_016857199.1
8, 9
885
XM_017001711.1
XP_016857200.1
8, 9
587
XM_017001712.1
XP_016857201.1
8, 9
885
XM_017001713.1
XP_016857202.1
6, 7
837
XM_017001714.1
XP_016857203.1
8, 9
871
XM_017001715.1
XP_016857204.1
8, 9
871
XM_017001716.1
XP_016857205.1
7, 8
739
XM_017001717.1
XP_016857206.1
7, 8
1060
XM_017001718.1
XP_016857207.1
6, 7
714
XM_017001719.1
XP_016857208.1
6, 7
812
XM_017001720.1
XP_016857209.1
6, 7
812
GenBank mRNA
AB209189.1
-
9, 10
865
AB451239.1
-
6, 7
544
AB451364.1
-
6, 7
544
AK091420.1
-
7, 8
495
AK296482.1
-
9, 10
596
AK304322.1
-
6, 7
724
AK304375.1
-
6, 7
539
AK314550.1
-
6, 7
791
AK316115.1
-
5, 6
506
BC016285.1
-
6, 7
726
3916100
BC035058.1
-
6, 7
813
5262372
BI598037.1
-
6, 7
610
BM905428.1
-
6, 7
681
BX537705.1
-
6, 7
776
BX641026.1
-
6, 7
775
BX647515.1
-
9, 10
874
CR936631.1
-
8, 9
588
DA228647.1
-
5, 6
534
M34181.1
-
6, 7
591
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00176944_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-kd7lp:80/10.239.160.160:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0