Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 17 | 1968 | ||
GenBank mRNA | AF548565.1 | - | 17 | 1911 | ||
BC022588.1 | - | 2 | 784 | |||
BC046963.1 | - | 17 | 1906 | |||
BC100666.1 | - | 12 | 1274 |