Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026250.2 | NP_080526.2 | 2 | 196 | ||
GenBank mRNA | AK014287.1 | - | 2 | 208 | ||
AK015243.1 | - | 2 | 175 | |||
AK018404.1 | - | 2 | 197 | |||
AK084620.1 | - | 1 | 100 | |||
AK090002.1 | - | 2 | 181 | |||
AK169361.1 | - | 2 | 159 | |||
BC033572.1 | - | 1 | 141 | 5067692 |