Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Olfr987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011785.1 | NP_001011785.1 | 1 | 791 | ||
GenBank mRNA | BC147165.1 | - | 1 | 867 | 9056167 | |
BC147166.1 | - | 1 | 867 | 9056185 |
Gene Symbol: Olfr988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011534.1 | NP_001011534.1 | 1 | 791 | ||
GenBank mRNA | BC150840.1 | - | 1 | 865 | 9087751 |