Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11163
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
7
793
NM_001318068.1
NP_001304997.1
7
888
NM_001318069.1
NP_001304998.1
8
1358
NM_002753.4
NP_002744.1
8
894
NM_138980.3
NP_620446.1
8
1385
NM_138982.3
NP_620448.1
8
1358
XM_005263129.2
XP_005263186.1
8
1455
XM_005263130.2
XP_005263187.1
8
851
XM_005263131.2
XP_005263188.1
8
1252
XM_005263135.3
XP_005263192.1
8
1252
XM_006714268.2
XP_006714331.1
8
1125
XM_006714269.2
XP_006714332.1
8
1252
XM_011532117.2
XP_011530419.1
8
958
XM_011532118.2
XP_011530420.1
8
1051
XM_011532120.2
XP_011530422.1
8
1021
XM_011532121.2
XP_011530423.1
8
887
XM_017008419.1
XP_016863908.1
9
1148
XM_017008420.1
XP_016863909.1
8
1582
XM_017008421.1
XP_016863910.1
8
1178
XM_017008422.1
XP_016863911.1
6
678
XM_017008423.1
XP_016863912.1
8
870
XM_017008424.1
XP_016863913.1
8
1205
XM_017008425.1
XP_016863914.1
8
878
XM_017008426.1
XP_016863915.1
8
1206
XM_017008427.1
XP_016863916.1
8
1279
XM_017008428.1
XP_016863917.1
8
1021
XM_017008429.1
XP_016863918.1
8
1582
XM_017008430.1
XP_016863919.1
8
960
XM_017008431.1
XP_016863920.1
8
1178
XM_017008432.1
XP_016863921.1
8
851
XM_017008433.1
XP_016863922.1
8
1279
XM_017008434.1
XP_016863923.1
8
1235
XM_017008435.1
XP_016863924.1
8
1125
XM_017008436.1
XP_016863925.1
8
1609
XM_017008437.1
XP_016863926.1
8
887
XM_017008438.1
XP_016863927.1
8
878
XM_017008439.1
XP_016863928.1
8
1206
XM_017008440.1
XP_016863929.1
8
1279
XM_017008441.1
XP_016863930.1
8
1279
XM_017008442.1
XP_016863931.1
10
1600
XM_017008443.1
XP_016863932.1
10
1251
XM_017008444.1
XP_016863933.1
10
1573
XM_017008445.1
XP_016863934.1
7
1082
XM_017008446.1
XP_016863935.1
5
1774
XM_017008447.1
XP_016863936.1
7
1220
XM_017008448.1
XP_016863937.1
7
1193
XM_017008449.1
XP_016863938.1
7
1220
XM_017008450.1
XP_016863939.1
7
1193
XM_017008451.1
XP_016863940.1
7
1019
XM_017008452.1
XP_016863941.1
7
1220
XM_017008453.1
XP_016863942.1
6
678
XM_017008454.1
XP_016863943.1
8
958
GenBank mRNA
AK057723.1
-
3
680
AK124791.1
-
8
1610
AK225974.1
-
8
939
AK294125.1
-
4
409
BC022492.1
-
8
1191
BC035057.1
-
8
1203
5261221
BC051731.1
-
8
1004
4838495
BC065516.1
-
3
355
5721954
U07620.1
-
8
895
U34819.1
-
7
739
U34820.1
-
7
763
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-wz2wq:80/10.239.167.51:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0