Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr1231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146454.2 | NP_666665.2 | 1 | 88 | ||
GenBank mRNA | BC104282.1 | - | 1 | 84 | ||
BC104283.1 | - | 1 | 87 |