Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Olfr679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147044.1 | NP_667255.1 | 1 | 662 | ||
GenBank mRNA | BC127993.1 | - | 1 | 666 | 40135720 |
Gene Symbol: Olfr681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207557.2 | NP_997440.2 | 1 | 662 | ||
GenBank mRNA | BC104298.1 | - | 1 | 662 | 40043142 | |
BC104299.1 | - | 1 | 662 | 40043148 |