Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172392.3 | NP_765980.3 | 4 | 984 | ||
GenBank mRNA | AK030126.1 | - | 4 | 987 | ||
BC028265.1 | - | 4 | 968 | 5365910 |