Search Thermo Fisher Scientific
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024645.1 | NP_001019816.1 | 29, 30 | 2852 | ||
GenBank mRNA | AK131164.1 | - | 29, 30 | 2795 | ||
BC060252.1 | - | 10, 11 | 1392 |