Search
Search

Cpsf7| Gene Symbol: Cpsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164272.1 | NP_001157744.1 | 2, 3 | 202 | ||
| NM_172302.3 | NP_758506.3 | 2, 3 | 202 | |||
| GenBank mRNA | AK044306.1 | - | 2, 3 | 181 | ||
| AK053375.1 | - | 2, 3 | 181 | |||
| AK088603.1 | - | 2, 3 | 180 | |||
| AK165330.1 | - | 2, 3 | 180 | |||
| AK188327.1 | - | 1, 2 | 70 | |||
| AK189206.1 | - | 1, 2 | 70 | |||
| BC017519.1 | - | 1, 2 | 122 | |||
| BC038812.1 | - | 1, 2 | 132 | 4506179 | ||